VWR® Autoclavable PE Biohazard Bags, 4 mil
Supplier: VWR International
11215-824 11215-822 11215-823
11215-824CS 1127.25 CAD
11215-824 11215-822 11215-823
VWR® Autoclavable PE Biohazard Bags, 4 mil
Bags Waste Disposal Bags
These heavy-duty bags with extra puncture resistance and high impact strength are constructed of high-molecular, high-density 4 mm polyethylene plastic.
  • Bright red color for quick identification
  • Clearly labeled with biohazard warnings
Autoclavable up to 121 °C (250 °F). Ideal for sterilizing animal cages. Each package includes 20.3 cm (8") twist ties. Bags meet many state and local regulations.
Order Now
SPECIFICATIONS
Environmentally Preferable