VWR® Adjustable Spray Bottle
Supplier: VWR
VWR®
3345
23609-182PK 82.63 CAD
23609-182
VWR® Adjustable Spray Bottle
Bottles Spray Bottles
The bottle's adjustable sprayer permits a range of positioning from fine mist to large stream delivery to accommodate a variety of washing applications.
  • HDPE and polypropylene construction
  • Chemical resistant and unbreakable
  • Instant-off release eliminates messy dripping
The chemical-resistant and unbreakable bottle is constructed of HDPE and the sprayer is made from polypropylene. The pump trigger is comfortable for one-handed operation and features an instant-off release that eliminates messy dripping. Closed position on sprayer maintains solution purity. The entire wash bottle may be gas or chemically sterilized. Ideal for hot, cold, aqueous, and solvent-based solutions.