Nalgene® Unitary™ Vented Safety Wash Bottles, Low-Density Polyethylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
2436-0501 2436-0502 2436-0503 2436-0504 2436-0505 2436-0506 2436-1001 2436-1005
16210-915CS 489.29 CAD
16210-915 16210-917 16210-919 16210-921 16210-923 16210-925 16210-927 16210-935
Nalgene® Unitary™ Vented Safety Wash Bottles, Low-Density Polyethylene, Thermo Scientific
Bottles Wash Bottles
LDPE bottles feature a unique vent system to keep hazardous volatile liquids in the bottle and off the lab bench.
Sodium hypochlorite (bleach) bottles are supplied with vented, white HDPE 38 mm closure; all other bottles supplied with vented polypropylene 38 mm colored closure. To prevent cross-contamination, each bottle has a color bar below the chemical name that corresponds to the colored closure.
Unitary design prevents contamination when filling the bottle. A strong, steady stream of liquid is dispensed when the bottle is squeezed.