Trigger Spray Bottles, Qorpak
Supplier: Qorpak
PLC-06934 PLC-03431 PLC-03499 PLC-03438 PLC-07581
11311-616CS 82.72 CAD
11311-616 16344-101 16344-508 93001-660 76487-100
Trigger Spray Bottles, Qorpak
Bottles Spray Bottles
Trigger spray bottles feature a trigger-motion spray that dispenses a spray or stream of liquid from an adjustable nozzle.
The translucent HDPE bottles are ideal for general use. HDPE provides an excellent moisture barrier and impact resistance. The adjustable sprayer permits a versatile spray pattern, from fine mist to coarse spray to jet stream.