Chemware® Wash Bottles, Teflon® PFA, Wide Mouth, Saint-Gobain
Supplier: Saint Gobain Performance Plastics
D1069739 D1069740 D1069741 D1069742
16651-880EA 400.72 CAD
16651-880 16651-881 16651-882 16651-883
Chemware® Wash Bottles, Teflon® PFA, Wide Mouth, Saint-Gobain
Bottles Wash Bottles
Transparent and unbreakable bottles can be rinsed with or used to dispense virtually all chemicals.
PFA fluoropolymer material withstands temperatures to 500 °F. Bottles are chemically inert, non-stick and easy to clean. Flex life and mechanical strength is superior to FEP. PFA contains the lowest concentration of trace metal ions of all fluoropolymers.