Nalgene® Economy Wash Bottles, Low-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
2401-0125 2401-0250 2401-0500 2401-1000
16651-450CS 334.14 CAD
16651-450 16651-471 16651-493 16651-518
Nalgene® Economy Wash Bottles, Low-Density Polyethylene, Narrow Mouth, Thermo Scientific
Bottles Wash Bottles
Through-the-top wash bottles feature closure and stem molded in one piece for leakproof service.
Draw tube is welded into closure to prevent separation. The tip dispenses a uniform stream, and can be cut back to increase flow. Supplied with polypropylene screw cap and stem.
Caution: Not recommended for long-term use with acetone.