Nalgene® Right-To-Know Safety Wash Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
2425-0501 2425-0502 2425-1002 2425-0503 2425-1003 2425-0504 2425-1004 2425-0505 2425-1005 2425-0500 2425-0506
16651-965CS 553.16 CAD
16651-965 16651-970 16651-972 16651-975 16651-977 16651-980 16651-982 16651-985 16651-987 16651-991 16652-020
Nalgene® Right-To-Know Safety Wash Bottles, Thermo Scientific
Bottles Wash Bottles
Wash bottles are silk screened with high-quality, chemical-resistant inks.
To prevent cross-contamination, each bottle has a color bar that corresponds to its colored closure for easy hazard and chemical identification. These leakproof bottles meet requirements of OSHA Hazard Communication Regulation 29 CFR 1910.1200(f)(4).
All bottles except sodium hypochlorite bottles are molded of translucent LDPE for chemical resistance and durability, and include a polypropylene closure. Sodium hypochlorite bottles are molded of white LDPE for light-protective barrier qualities between 280 and 450 nm. They include an HDPE closure.
Ordering information: Assortment 16651-991 includes bottles (500 ml) and closures for all chemicals listed except sodium hypochlorite.