Breather Bags, Sheeting and Tubing, KNF
Supplier: KNF Cleanroom Products
600037-46 600037-59 600105-01 600105-07 600076-04 600076-07 600076-08 600076-09 600076-10
12243-240CS 214.74 CAD
12243-240 12243-246 12243-252 12243-262 12243-272 12243-276 12243-278 12243-836 12243-838 12243-840 12243-842 12243-844
Breather Bags, Sheeting and Tubing, KNF
Bags Sterilization Pouches
Ideal for use in pharmaceutical and medical industries where FDA Class 100 sterilization is required.
Bags are made from two sheets of copolymer polypropylene with a strip of Tyvek® 1059B heat-sealed over the slits on one of the sheets.
When subjected to varying chamber pressures, the breather slits help equalize internal and external pressures, while preventing product from contacting the outer Tyvek® barrier. Breather slits are 10.16 cm (4") wide and located at the center of bag or sheet. When sizes cannot be predetermined, Breather Tubing is also available. Thickness: 3 mil.
Sterilizable and disposable, these products can withstand temperatures up to 128 °C for extended periods and remain exceptionally pliable.
Certifications: Cleanliness level 1.
Packaging: Supplied triple-barrier packaged with nylon inner bag and two polyethylene outer bags.