VWR® Red Wash Bottles, LDPE
Supplier: VWR International
11215-302 16650-537
11215-302BG 79.47 CAD
11215-302 16650-537
VWR® Red Wash Bottles, LDPE
Bottles Wash Bottles
Low-density polyethylene bottles molded in bright red provide easy identification of hazardous materials.
  • Vibrant red color for quick identification
  • Ideal for dangerous solvents and solutions
Durable, flexible, chemical-resistant, and easy to clean. Excellent pliability allows comfortable squeeze flow control. Has one-piece molded polypropylene cap and nozzle. Dispense a single drop or a needle sharp stream. If a coarser flow is desired, dispensing tip can be trimmed.
Order Now
SPECIFICATIONS
Environmentally Preferable