Kapak SealPAK 400 and 500 Series Pouches, Ampac Flexibles
Supplier: Ampac Flexibles
502B-1M 402B-2M 404B-2M 400B-2M 500B-1M 504B-1M 505B-1M 506B-500 508-24 400-24 402-24 403-24 404-24 406-24 405-24 500-24 502-24 503-24 504-24 506-24 505-24
11213-916CS 2180.37 CAD
11213-916 11214-560 11214-562 11214-564 82007-176 82007-178 82007-180 82007-182 11213-918 11214-301 11214-334 11214-338 11214-367 11214-390 11214-394 11214-469 11214-491 11214-495 11214-527 11214-550 82017-786
Kapak SealPAK 400 and 500 Series Pouches, Ampac Flexibles
Bags Sample Bags
Boilable, freezable, and microwavable polyester/polyethylene (PET/PE) laminated Kapak pouches combine tensile strength with strong heat seals and superior barrier properties.
Regular weight 2.5 mil material is best for moderate protection from handling and chemical or environmental contact. Tear notches included. Heavy-duty 4.5 mil material is best for institutional and industrial use; they are also suitable for long-term storage and mailing. Tear notches not included.
These pouches will sustain temperatures from –56 to +115 °C (–70 to +240 °F). Safe for use with concentrations of formaldehyde, most acids, alkalies, greases, oils, and organic solvents. Transparent, airtight, moisture, and odor-proof. They can withstand infrared and gamma rays. These pouches can be surface sterilized using VHP. Ideal for histology and pathology specimens, as well as safe biological waste disposal in medical, institutional, and industrial facilities. Seal using vacuum or commercial heat-sealing equipment