VWR® Safety-Vented™ Wide Mouth Labeled Wash Bottles, LDPE
Supplier: VWR International
66001-148 66001-152 89141-044 89141-048 CA66001-141 CA66001-155
66001-148PK 76.88 CAD
66001-148 66001-152 89141-044 89141-048 CA66001-141 CA66001-155
VWR® Safety-Vented™ Wide Mouth Labeled Wash Bottles, LDPE
Bottles Wash Bottles
Wide mouth, color-coded, low-density polyethylene wash bottles feature a unique two-piece polypropylene venting assembly that prevents pressure buildup of solvents inside the bottle.
  • Pre-labeled and vented for maximum safety
  • Eliminate solvent drips
  • Easy-to-read labeling
Venting assembly eliminates solvent drips from the spout due to internal evaporation. Easy-to-read solvent labeling includes DOT, NFPA codes and symbols, as well as CAS numbers. Bottles are leakproof.
Certifications: Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
Caution: LDPE wash bottles are intended for short-term dispensing use only and not for long-term storage. Bottles marked LYOB (Label Your Own Bottle) are designed for use only with solvents compatible with LDPE.