KIMAX® Solution Bottles with Color-Coded PTFE [ST] Flathead Stopper, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
15097-100 15097-1000 15097-2000 15097-250 15097-500
89001-576CS 161.42 CAD
89001-576 89001-578 89001-580 89001-582 89001-584
KIMAX® Solution Bottles with Color-Coded PTFE [ST] Flathead Stopper, Kimble Chase, DWK Life Sciences
Bottles Solution Bottles
Narrow-mouth bottles conform to ASTM E438, Type 1, Class A standards.
Bottle necks are [ST] ground to accept flathead PTFE stoppers.