VWR® Sterile Sample Bags with Specimen Sponge
Supplier: VWR
VWR®
KSS-61105 KSS-61100 KSS-61100-VW1 KSS-61105
CA89085-576CS 3586 CAD
89003-806 CA89085-576 89085-576 CA89003-806
VWR® Sterile Sample Bags with Specimen Sponge
Bags Sample Bags
Sterile sample bags are ideal for surface sampling in a wide range of applications.
  • Sterile
  • Includes cellulose sponge
  • Polyethylene sterile tubing
  • Write-on strip
  • Wire closure for an airtight seal
  • Wide-sealed bottoms for leakproof protection
They provide a reliable method for monitoring potentially hazardous environments for the presence of microorganisms as well as foodborne pathogens.
Each bag includes a cellulose sponge that measures 3.8Wx7.6Lx1.6Hcm (11/2x3x5/8") when hydrated. Choose from a biocide-free, dry sponge or a foil-sealed sponge premoistened with a sterile buffer solution.
Bags are made of virgin polyethylene tubing with no side seals. They feature a wire closure for an airtight seal, wide-sealed bottoms for added leakproof protection, and a write-on strip. They are easy to use: tear off perforated top, pull tabs to open bag, fill, grab ends of tape closure and pull shut, twirl bag three or four times to seal, and bend ends of closure inward to lock the seal.
Caution: Do not use with alcoholic substances or where temperature exceeds 100°C (212°F).