VWR® Wide Mouth Wash Bottles, LDPE
Supplier: VWR International
16650-082 16650-093 16650-107 16650-162 16650-173 16650-184 16650-195 16650-242 16650-264 16650-322 16650-333 16650-344 16650-355
16650-082CS 660.47 CAD
16650-082 16650-093 16650-107 16650-162 16650-173 16650-184 16650-195 16650-242 16650-264 16650-322 16650-333 16650-344 16650-355
VWR® Wide Mouth Wash Bottles, LDPE
Bottles Wash Bottles
These bottles save time, help avoid mistakes, and increase safety.
  • Color-coded caps provide quick, safe identification
  • Wide-mouthed design
  • One-piece cap and nozzle plus delivery tube
  • Single drop or needle-thin stream
Color-coded, wide mouth bottles are ideal for the quick identification of the many special solutions and reagents needed in your lab. Bottles are durable, chemical-resistant, squeezable, and easy to clean. One-piece colored cap and nozzle are polypropylene. Delivery tube is polyethylene. The leakproof wash cap assembly will deliver aMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable