MechPro Utility Gloves, Wells Lamont
Supplier: Wells Lamont
7701S 7701M 7701L 7701XL Y7711S Y7711M Y7711L Y7711XL
83007-986PK 541.28 CAD
83007-986 83007-988 83007-990 83007-992 83007-994 83007-996 83007-998 83008-002
MechPro Utility Gloves, Wells Lamont
Gloves
MechPro plus gloves have all the features of MechPro standard gloves and additional padding in the palm area to reduce bruising and damage from vibration and excessive grip pressure.
  • Padding minimizes Ssock
  • Heat-resistant knit fabric
  • Reinforced fingertips
Padding helps to minimize shock, which can lead to carpal tunnel syndrome.
Gusseted finger crotches provide additional comfort and flexibility. Gloves are double-stitched in high wear areas: fingertips, index fingers, and palms. An elastic cuff with hook and loop closure provides a secure fit that keeps out debris.
Heavy-duty, thick-ribbed, knit fabric back provides protection from nicks and abrasions and resists incidental heat. A synthetic leather palm and fingertips also offer protection while enhancing grip. Reinforced fingertips feature pinched seams to increase tactile sensitivity.