Anti-Static Poly Zipper Bags, Associated Bag
Supplier: Associated Bag
104-6-33 103-07
11215-934CS 563.54 CAD
11215-934 47743-538
Anti-Static Poly Zipper Bags, Associated Bag
Bags Sample Bags
Side-seal construction and reinforced spot-welded zipper protect sensitive items from damage caused by static electricity, dirt, and moisture.
Resealable, single-track zipper stands up to repeated use. Transparent bags with pink tint are easily identified during shipping or storage. Manufactured to meet the static decay requirements of Mil-PRF-81705E. Usable dimensions are slightly smaller.