Potassium hydrogen carbonate ≥99.5% (dry basis), powder USP, FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Potassium bicarbonate

Management of Change (MOC) category = R

MACR6736-04 MACR6736-44
CAMK673604CS 2197.32 CAD
CAMK673604 CAMK6736-44
Potassium hydrogen carbonate ≥99.5% (dry basis), powder USP, FCC, Macron Fine Chemicals™
Potassium hydrogen carbonate

Hygroscopic powder.


  • Food GMP manufactured product

Formula: KHCO₃
MW: 100.12 g/mol
Melting Pt: 292
Density: 2.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011402
CAS Number: 298-14-6
EINECS: 206-059-0
Merck Index: 13,07691

Order Now

Specification Test Results

Meets F.C.C. Requirements
Meets U.S.P Requirements
GMP Manufactured Product
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Assay (KHCO₃) (dried basis) 99.5 - 101.5 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Loss on Drying ≤0.25 %
USP - Normal Carbonate ≤2.5 %
FCC - Assay (KHCO₃) (dried basis) 99.5 - 101.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Carbonate Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Loss on Drying ≤0.25 %
Solubility Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR