Potassium hydrogen phthalate ≥99.95%, crystals, AR® ACS primary standard, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt

Management of Change (MOC) category = N

6704-02 6704-04 MACR6704-04
MK670402CS 4117.15 CAD
MK670402 MK670404 CAMK670402 CAMK670404 CAMK670420
Potassium hydrogen phthalate ≥99.95%, crystals, AR® ACS primary standard, Macron Fine Chemicals™
Potassium hydrogen phthalate

Primary standard.

Formula: HOOCC₆H₄COOK
Melting Pt: 295…300 °C
Density: 1.6362 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013070
CAS Number: 877-24-7
EINECS: 212-889-4
Merck Index: 13,07694
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4652479e822d1591ac7eb5e29f892b8c94588ad4&vid=75071da7f30b07f7bd077699a3b54ec89d80f0b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (1-KOCOC₆H₄-2-COOH) 99.95 - 100.05 %
ACS - Chlorine Compounds (as Cl) ≤0.003 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤5 ppm
ACS - pH of 0.05M Solution at 25°C 4.00 - 4.02
ACS - Sodium (Na) ≤0.005 %
ACS - Sulfur Compounds (as S) ≤0.002 %
Traceable to NIST
Appearance (white crystals)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR