Orthophosphoric acid ≥85.0-88.0% (by acidimetry) NF, FCC for biochemistry, suitable for use as excipient, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Phosphoric acid

Management of Change (MOC) category = R

MACR2788-46 MACR2788-14 MACR2788-26 MACR2788-27
CAMK278846CS 2902.03 CAD
CAMK278846 CAMK278814 CAMK278826 CAMK2788-27
Orthophosphoric acid ≥85.0-88.0% (by acidimetry) NF, FCC for biochemistry, suitable for use as excipient, Macron Fine Chemicals™
Orthophosphoric acid

pH adjusters are used to adjust the pH of formulations. This category includes acids that are notoriously difficult to source as truly cGMP-produced, and amino acids that most companies do not fully test for EP, BP and JP. Avantor provides all that and more with our cGMP-produced acids and mutli-compendial amino acids.

Formula: H₃PO₄
MW: 98 g/mol
Boiling Pt: 158
Melting Pt: 21
Density: 1.71…1.87 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8 III
Merck Index: 14,07344

Order Now

Specification Test Results

GMP Manufactured Product
Meets F.C.C. Requirements
Food GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
NF - Assay (H₃PO₄) (by acidimetry) 85.0 - 88.0 %
NF - Alkali Phosphates Passes Test
NF - Identification Passes Test
NF - Nitrate (NO₃) Passes Test
NF - Phosphorous or Hypophosphorous Acid Passes Test
NF - Sulfate (SO₄) Passes Test
FCC - Assay (H₃PO₄) (by acidimetry) 85.0 - 88.0 %
FCC - Arsenic (As) ≤3 mg/kg
FCC - Cadmium (Cd) ≤3 mg/kg
FCC - Fluoride (F) ≤10 mg/kg
FCC - Identification Passes Test
FCC - Lead (Pb) ≤3 mg/kg
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Appearance (clear, colorless solution)
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR