2-Propanol 99,5, ChromAR® LC, UV, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

M.O.C. Category N

3043-06 MACR3043-10 MACR3043-23
MK304306CS 1440.91 CAD
MK304306 CAMK304310 CAMK304323
2-Propanol 99,5, ChromAR® LC, UV, Macron Fine Chemicals™
2-Propanol

ChromAR®—Solvents that meet ACS specifications and are suitable for liquid chromatography and UV-spectrophotometry. 

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃CHOHCH₃) ≥99.5 %
Carbonyl Compounds - Propionaldehyde ≤0.002 %
Carbonyl Compounds - Acetone ≤0.002 %
Color (APHA) ≤10
Residue after Evaporation ≤0.0005 %
Solubility in H₂O Passes Test
Titrable Acid or Base (meq/g) ≤0.0001
Water (H₂O)(by Karl Fischer titrn) ≤0.05 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 205 nm ≤1.00
Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm ≤1.00
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.30
Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm ≤0.15
Ultraviolet Absorbance (1.00-cm cell vs. water) - 245 nm ≤0.08
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm ≤0.02
Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm ≤0.04
Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm ≤0.03
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm ≤0.02
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 - 330 nm ≤0.01
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR