Perchloric acid 69,0 - 72,0% ≥70%, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

MACR2766-44 MACR2766-14
CAMK2766-44CS 3485.72 CAD
CAMK2766-44 CAMK276614
Perchloric acid 69,0 - 72,0% ≥70%, AR® ACS, Macron Fine Chemicals™
Perchloric acid

ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.  

Formula: HClO₄
Boiling Pt: 160…181 °C (1013 hPa)
Melting Pt: –112.2 °C
Density: 1.53…1.768 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (HClO₄) 69.0 - 72.0 %
Appearance (clear, colorless solution) Passes Test
ACS - Chloride (Cl) ≤ 0.001 %
ACS - Color (APHA) ≤ 10
ACS - Heavy Metals (by ICP-OES) ≤ 1 ppm
ACS - Iron (Fe) ≤ 0.0001 %
ACS - Nitrogen Compounds (as N) ≤ 0.001 %
ACS - Residue after Ignition ≤ 0.003 %
ACS - Silicate and Phosphate (as SiO₂) ≤ 0.0005 %
ACS - Sulfate (SO₄) ≤ 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR