Potassium chloride 99.0-100.5% USP, FCC, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
|
MACR6838-20
MACR6838-26
CAMK683820EA
2695.91
CAD
CAMK683820
CAMK683826
Potassium chloride 99.0-100.5% USP, FCC, Macron Fine Chemicals™
Potassium chloride
Granular. GMP manufactured.
Formula:
KCl Boiling Pt: 1420 °C (1013 hPa) Melting Pt: 773 °C Density: 1.98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011360 CAS Number: 7447-40-7 EINECS: 231-211-8 Merck Index: 14,07621 |
Specification Test Results
Meets U.S.P Requirements | |
Meets F.C.C. Requirements | |
GMP Manufactured Product | |
Food GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Food Chemical | |
Bulk Pharmaceutical Chemical | |
USP - Assay (KCl) (by Ag titrn) | 99.0 - 100.5 % |
USP - Acidity or Alkalinity (no pink color then pink) | Passes Test |
USP - Bromide (Br)(USP) | ≤0.1 % |
USP - Calcium and Magnesium | Passes Test |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Iodide (I)(USP) | ≤0.005 % |
USP - Loss on Drying at 105°C | ≤1.0 % |
USP - Sodium (Na) | Passes Test |
FCC - Assay (KCl) (by Ag titrn) | 99.0 - 100.5 % |
FCC - Acidity or Alkalinity (no pink color then pink) | Passes Test |
FCC - Bromide and/or Iodide (FCC) | Passes Test |
FCC - Heavy Metals (as Pb)(FCC) | ≤5 mg/kg |
FCC - Identification A | Passes Test |
FCC - Identification B | Passes Test |
FCC - Loss on Drying at 105°C | ≤1.0 % |
FCC - Sodium (Na) | Passes Test |
Solubility (10 in 50) | Passes Test |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...