Potassium hydroxide 85.0-100.5%, pellets NF, FCC, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = R
6976-28
MACR6976-06
MACR6976-25
MK697628EA
2777.26
CAD
MK697628
CAMK697606
CAJT6976-QM
CAMK697620
CAMK697625
Potassium hydroxide 85.0-100.5%, pellets NF, FCC, Macron Fine Chemicals™
Potassium hydroxide
Pellets. GMP manufactured.
Formula:
KOH Boiling Pt: 1320 °C (1013 hPa) Melting Pt: 360 °C Density: 2.04 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 EINECS: 215-181-3 UN: 1813 ADR: 8,II REACH: 01-2119487136-33 Merck Index: 12,07806 |
Specification Test Results
Meets F.C.C. Requirements | |
Meets N.F. Requirements | |
GMP Manufactured Product | |
Food GMP Manufactured Product | |
Storage: Preserve in tight containers. | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
Bulk Food Chemical | |
NF - Assay (Total Alkali) | ≥85.0 % |
NF - Carbonate (as K₂CO₃) | ≤3.5 % |
NF - Content of Potassium | ≥59.9 % |
NF - Limit of Sodium | ≤1.0 % |
NF - Identification A | Passes Test |
NF - Identification B | Passes Test |
NF - Insoluble Substances | Passes Test |
FCC - Assay (KOH) | 85.0 - 100.5 % |
FCC - Identification A | Passes Test |
FCC - Identification B | Passes Test |
FCC - Insoluble Substances | Passes Test |
FCC - Lead (Pb) | ≤2 mg/kg |
FCC - Mercury (Hg) | ≤0.1 ppm |
FCC - Potassium Carbonate (K₂CO₃) | ≤3.5 % |
Solubility | Passes Test |
Appearance (white hygroscopic pellets) | |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...