Potassium peroxodisulfate ≥99.0%, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

7076-12 MACR7076-20
MK707612CS 1085.54 CAD
MK707612 CAMK707620
Potassium peroxodisulfate ≥99.0%, AR® ACS, Macron Fine Chemicals™
Potassium peroxodisulphate

ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.  

Formula: K₂S₂O₈
Melting Pt: 100 °C
Density: 2.477 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011386
CAS Number: 7727-21-1
EINECS: 231-781-8
UN: 1492
ADR: 5.1,III
Merck Index: 13,07741

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (K₂S₂O₈) ≥99.0 %
ACS - Chlorine Compounds (as Cl) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤0.0005 %
ACS - Manganese (Mn) ≤0.0002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR