1-Propanol 99,0 GC, corrected for water content, AR®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane

M.O.C. Category N

MACR7169-04 MACR7169-19
CAMK716904CS 1582.81 CAD
CAMK716904 CAMK716919 CAMK716908
1-Propanol 99,0 GC, corrected for water content, AR®, Macron Fine Chemicals™
1-Propanol
Formula: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Boiling Pt: 97
Melting Pt: -127
Density: 0.8053 g/cm³ (20 °C)
Flash Pt: 15
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3 II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃(CH₂)₂OH) (by GC, corrected for water) ≥99.0 %
Acidity (as CH₃CH₂COOH) ≤0.015 %
Alkalinity (as NH₃) ≤0.002 %
Residue after Evaporation ≤0.0050 %
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR