Whatman™ Reaction Folders - Sealable Hybridization Bags, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10483064 10483064
CA89027-004PK 189.52 CAD
CA89027-004 89027-004
Whatman™ Reaction Folders - Sealable Hybridization Bags, Whatman products (Cytiva)
Bags Roll Stock
Whatman reaction folders are sealable bags for hybridization and incubation reactions.
  • Folders can be sealed with a standard heat-sealer
The folders are open on three sides to allow easier insertion of a wet membrane when compared to standard hybridization bags.