Polyethylene glycol 4,000, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: PEG 4,000PEG 4000

Management of Change (MOC) category = N

H273-61 MACRH273-61
CAMKH27361EA 482.55 CAD
CAMKH27361 MKH27361
Polyethylene glycol 4,000, Macron Fine Chemicals™
Polyethylene glycol 4,000

Powder. Infrared spectrum conforms to reference standard.

Formula: H(OCH₂CH₂)nOH
MW: 4000 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 53…58 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 250 °C
MDL Number: MFCD01779612
CAS Number: 25322-68-3

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Color (APHA) ≤25
Infrared Spectrum Passes Test
Melting Range(Lower) 54 - 60 °C
Melting Range(Upper) 54 - 60 °C
Appearance (white powder or creamy-white flakes)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR