Phenolphthalein 1.0% in 50% alcohol, StandARd®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

Management of Change (MOC) category = N

H295-05 MACRH295-05
CAMKH29505EA 146.4 CAD
CAMKH29505 MKH29505
Phenolphthalein 1.0% in 50% alcohol, StandARd®, Macron Fine Chemicals™
Phenolphthalein
Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
Density: 0,92 Kg (1 L)
Flash Pt: 13 °C
MDL Number: MFCD00005913
CAS Number: 77-09-8
UN: 1993
ADR: 3,II
https://ca.vwr.com/stibo/web/std.lang.all/72/01/2297201.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
pH ≥8.0
pH ≥10.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR