Potassium chloride saturated aqueous solution, StandARd®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Management of Change (MOC) category = N
MACRH300-01
MACRH300-05
CAMKH30001EA
105.73
CAD
CAMKH30001
CAMKH30005
Potassium chloride saturated aqueous solution, StandARd®, Macron Fine Chemicals™
Potassium chloride
For refilling calomel reference electrodes.
Formula:
KCl Density: 1.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011360 CAS Number: 7447-40-7 EINECS: 231-211-8 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Density (g/mL) at 20°C | 1.162 - 1.172 |
Identification | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...