Perchloric acid 0.10 N in glacial acetic acid, StandARd® volumetric solution, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

MACRH301-05 MACRH301-07
CAMKH30105EA 291.48 CAD
CAMKH30105 CAMKH30107
Perchloric acid 0.10 N in glacial acetic acid, StandARd® volumetric solution, Macron Fine Chemicals™
Perchloric acid
Formula: HClO₄
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 2920
ADR: 8 (3),II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.0995 - 0.1005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR