Phenol solution, StandARd®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = N
MACRH302-01
CAMKH30201EA
48.35
CAD
CAMKH30201
Phenol solution, StandARd®, Macron Fine Chemicals™
Phenol
Formula:
C₆H₅OH |
MDL Number:
MFCD00002143 CAS Number: 108-95-2 UN: 1671 ADR: 6.1,II |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Concentration (0.8-1.2 mg/mL) | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...