Phenylarsine oxide 0.00564N Stabilised zA

Supplier: Ricca Chemical

Synonyms: PAO, Oxophenylarsine, Arzene

R5750000-1C R5750000-4C
RC5750-32EA 385.52 CAD
RC5750-32 RC57501
Phenylarsine oxide 0.00564N Stabilised zA
Phenylarsine oxide

Phenylarsine Oxide (PAO) Titrant (APHA and ASTM)


  • 1mL = 0.2 mg chloride
  • Stabilized

Formula: C₆H₅AsO
MW: 168.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001990
CAS Number: 637-03-6

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.005640 ± 0.000005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR