Orthophosphoric acid 10% (v/v) in aqueous solution (1 + 9) analytical reagent

Supplier: Ricca Chemical
Warning

Synonyms: Phosphoric acid

R5850000-4A R5850000-500A R5850000-1A
RC58501EA 385.52 CAD
RC58501 RC585016 RC585032
Orthophosphoric acid 10% (v/v) in aqueous solution (1 + 9) analytical reagent
Orthophosphoric acid

APHA for chlorine.

Formula: H₃PO₄
Density: ∼1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 10.00 ± 0.03 % (v/v)
Specific Gravity 1.075 ± 0.005 sp gr at 20 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR