Potassium hydroxide 10% (w/v) in aqueous solution
Supplier: Ricca Chemical
|
Danger
|
R6130000-500A
R6130000-120A
R6130000-250A
RC613016EA
110.6
CAD
RC613016
RC61304
RCR6130000250A
Potassium hydroxide 10% (w/v) in aqueous solution
Potassium hydroxide
Formula:
KOH Boiling Pt: ~100 °C Melting Pt: ~0 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 EINECS: 215-181-3 UN: 1814 ADR: 8,III |
Specification Test Results
Assay (vs. Sulfuric Acid/Phenolphthalein) | 9.94 - 10.06 % (w/v) |
Appearance | Colorless liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...