Potassium hydroxide 20% (w/v) in aqueous solution analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Potash caustic, Caustic potash

R6140000-500A R6140000-1A R6140000-250A
RC614016EA 121.66 CAD
RC614016 RC6140-32 RCR6140000250A
Potassium hydroxide 20% (w/v) in aqueous solution analytical reagent
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 19.94 - 20.06 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR