Potassium hydroxide 1.00 Normal, Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Potash caustic, Caustic potash
R6250000-4A
R6250000-1A
R6250000-20F
RC62501EA
338.12
CAD
RC62501
RC625032
RC62505
Potassium hydroxide 1.00 Normal, Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility
Potassium hydroxide
Standardized at 25°C against potassium acid phthalate whose certification is traceable to NIST.
Formula:
KOH Storage Temperature: Ambient |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 EINECS: 215-181-3 UN: 1814 ADR: 8,III |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 84) | 1.000 ± 0.001 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...