(±)-1,2-Propanediol ≥99.5% (dry basis), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane

Management of Change (MOC) category = N

BAKRU510-01 BAKRU510-07 BAKRU510-09
CAJTU510-1EA 2940.4 CAD
CAJTU510-1 CAJTU510-7 CAJTU510-9
(±)-1,2-Propanediol ≥99.5% (dry basis), BAKER ANALYZED® ACS, J.T.Baker®
Propylene glycol

Lot analysis on label.

Formula: CH₃CH(OH)CH₂OH
MW: 76.1 g/mol
Boiling Pt: 187.6 °C (1013 hPa)
Melting Pt: –59 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00064272
CAS Number: 57-55-6
EINECS: 200-338-0
Merck Index: 13,07947
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃CHOHCH₂OH) (corrected for water) ≥99.5 %
ACS - Color (APHA) ≤10
ACS - Residue after Ignition ≤0.005 %
ACS - Titrable Acid (meq/g) ≤0.0005
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.2 %
Chloride (Cl) ≤1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR