UniSafe™ Vented Wash Bottles, LDPE
Supplier: United Scientific Supplies
UN370071 UN370072 UN370073 UN370074 UN370077 UN370079
470354-640PK 60.91 CAD
470354-640 470354-642 470354-644 470354-646 470354-648 470354-650
UniSafe™ Vented Wash Bottles, LDPE
Bottles Wash Bottles
Easy to read wash bottles with NFPA diamond and health hazards.
  • Industrial quality low density polyethylene (LDPE) construction with long lasting strength, GHS compliant
  • Preprinted bottles include: chemical name and formula, NFPA diamond, health hazards, and CAS number
  • Polypropylene stem closure and draw tube
  • Safety vented and color-coded for quick solvent recognition
  • 4 color printing, 1000 ml (32 oz.) capacity
UniSafe™ vented wash bottles are made from industrial grade low-density polyethylene with polypropylene stem closure and venting assembly. They are pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment. Each bottle is compliant with GHS and OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200. Intended for use only with LDPE and PP compatible solvents.