BIOXP® Select Plasmid AMP
Supplier: TELESIS BIO
BX-SEL-PLSAMP-08
BX-SEL-PLSAMP-16
BX-SEL-PLSAMP-24
77461-294EA
1914.96
CAD
77461-294
77461-296
77461-298
BIOXP® Select Plasmid AMP
Nucleic Acid Reagents
Cloning Kits and Ligation
BioXp Select plasmid amplification kits contain reagents for isothermal amplification of plasmid DNA.
- Input: Plasmids ranging from 1.8 to 37 kb
- Yield: >10 µg/well
- Output format: Branched concatemers
- Compatible with BioXp 3250
Contains reagents necessary to generate at least 10 micrograms of amplified DNA starting from a sequence verified plasmid supplied by the end user.