BIOXP® Select mRNA
Supplier: TELESIS BIO
BX-MRNA-RSU-CC-48
BX-MRNA-RSM1-CC-08
BX-MRNA-RSM1-CC-24
BX-MRNA-RSM1-CC-48
BX-MRNA-RSU-CC-08
BX-MRNA-RSU-CC-24
77461-284EA
13787.71
CAD
77461-284
77461-274
77461-276
77461-278
77461-280
77461-282
BIOXP® Select mRNA
Nucleic Acid Reagents
RNA Synthesis and Transcription
The BioXp Select mRNA synthesis kit enables the synthesis of mRNA from an input DNA template.
- mRNA Length: 0.4 to 10 kb
- Yield: >50 µg/well
- Compatible with BioXp 3250
- Cap: CleanCap® AG (3'OMe)
- Modification: Pseudouridine
Contains reagents necessary to generate at least 10 micrograms of amplified DNA starting from a sequence verified plasmid supplied by the end user.