BIOXP® 9600 Select Plasmid AMP
Supplier: TELESIS BIO
BX9-SEL-PLSAMP-08 BX9-SEL-PLSAMP-16 BX9-SEL-PLSAMP-24 BX9-SEL-PLSAMP-48 BX9-SEL-PLSAMP-96
77461-264EA 1276.64 CAD
77461-264 77461-266 77461-268 77461-270 77461-272
BIOXP® 9600 Select Plasmid AMP
Nucleic Acid Reagents Cloning Kits and Ligation
BioXp Select plasmid amplification kits contain reagents for isothermal amplification of plasmid DNA.
  • Input: Plasmids ranging from 1.8 to 37 kb
  • Yield: >10 µg/well
  • Output format: Branched concatemers
  • Compatible with BioXp 9600
Contains reagents necessary to generate at least 10 micrograms of amplified DNA starting from a sequence verified plasmid supplied by the end user.
Caution: Branched concatemers need to be linearized prior to transfection. A recommended transfection protocol is included.