BIOXP® 9600 Select Clone and AMP
Supplier: TELESIS BIO
BX9-SEL-CLNAMP-08
BX9-SEL-CLNAMP-16
BX9-SEL-CLNAMP-24
BX9-SEL-CLNAMP-48
BX9-SEL-CLNAMP-96
77461-254EA
1914.96
CAD
77461-254
77461-256
77461-258
77461-260
77461-262
BIOXP® 9600 Select Clone and AMP
Nucleic Acid Reagents
Cloning Kits and Ligation
The Select DNA cloning and amplification kits generate a minimum of 10 µg of amplified DNA from fragments of 0.3 to 1.8 kb using the BioXp 9600.
- Accepted input: Telesis Bio DNA fragments, IDT gBlocks™, Twist Biosciences fragments
- Supports user-provided linearized vector
- Length: 0.3 to 1.8 kb
- Yield: >10 µg/well
- Compatible with BioXp 9600
The BioXp Select DNA cloning and amplification kit contains Gibson Assembly® and other reagents necessary to synthesize at least 10 µg of amplified DNA from fragments and linearized vectors supplied by the user.