BIOXP® 3250 NGS Kits for Genomes
Supplier: TELESIS BIO
BX-NGS-WGS-08 BX-NGS-WGS-24
77461-290EA 638.32 CAD
77461-290 77461-292
BIOXP® 3250 NGS Kits for Genomes
Nucleic Acid Reagents Next Generation Sequencing Reagents
Generate Illumina-compatible, adapter-ligated, NGS libraries for whole genome sequencing in a simple automated workflow using the BioXp 3250.
  • Input: 10 to 20 ng of genomic DNA
  • Compatible with Illumina sequencers
  • 15 minutes of hands on time
  • Compatible with BioXp 9600
The BioXp NGS library prep kit for whole genome sequencing provides the necessary reagents, when combined with customer-provided sequencing adapters, to generate Illumina-compatible NGS libraries for assessment of genomic DNA. This kit uses a streamlined, fully automated ligation-based library prep method. The input into the kit is 10 to 20 ng of purified plasmids in nuclease-free water, Tris-HCl pH 8.0, or Tris low EDTA (0.1 mM).
Delivery information: Not supplied with necessary sequencing adapters.