Gibson Assembly® HIFI 1-Step Kits and Master Mixes (2X)
Supplier: TELESIS BIO
GA1100-S GA1100-10 GA1100-10MM GA1100-50 GA1100-50MM GA1100-B05
10820-608EA 182 CAD
10820-608 10820-610 10820-612 10820-614 10820-616 77463-574
Gibson Assembly® HIFI 1-Step Kits and Master Mixes (2X)
Nucleic Acid Reagents Cloning Kits and Ligation
Generate seamless plasmid DNA or BAC clones without restriction digestion.
  • Maximum nunber of fragments: 5
  • Cloning efficiency: 90% Full-length clones
  • Insert size range: 500 to 32000 bps
  • Total construct size range: 100000 bps
  • Reaction Time: 60 minutes
The Gibson Assembly process begins by designing dsDNA fragments with 20 to 40 bp overlapping ends. For the Gibson Assembly HiFi 1-Step reaction, these DNA fragments are combined with a 2X Gibson Assembly® HiFi 1-Step Master Mix and incubated. This during this time, complementary ends are generated, annealed, and sealed. This this one-step, isothermal process can be performed in less than 60 minutes and results in a transfection or transformation ready, double stranded, fully ligated DNA construct. The GA 1-Step Master Mix (2X) contains a proprietary mixture of enzymes and reagents optimized to facilitate one-step assembly of double stranded DNA fragments. This mastermix includes a proof-reading polymerase that mediates junction repair resulting assembled constructs with low rates of junction errors and high sequence fidelity.
The Gibson Assembly method allows the insertion of one or more linear double stranded DNA fragments into a virtually any vector without the need to rely on compatible restriction sites. Gibson Assembly is significantly faster than traditional restriction enzyme digest-based cloning and proven for the cloning of both small and large double stranded DNA fragments. Using the Gibson Assembly® HiFi 1-Step Master Mix, the simultaneous cloning of one to five inserts with sizes ranging from 500 bp to 32 kb can be accomplished in a single tube, in a single step, using a robust isothermal reaction that yields cloning efficiencies of greater than 90%.
This product does not include the GA Positive Control found in the Gibson Assembly® HiFi 1-Step Kit. If you are new to Gibson Assembly or require a validated positive control for you experiments, you may want to consider catalog numbers 10820-608, 10820-610 or 10820-612.
Ordering information: Kits with 5, 10, or 50 reactions are available. All kits contains one vial of GA 1-Step Master Mix (2X) and one vial of GA Positive Control (2X) and a detailed instruction manual.