Pinacol ≥98.0%

Supplier: TCI America

D0691-25G D0691-500G D0691-100G
TCD0691-025GEA 75.84 CAD
TCD0691-025G TCD0691-500G TCD0691-100G
Pinacol ≥98.0%
Pinacol

CAS Number: 76-09-5
MDL Number: MFCD00004462
Molecular Formula: C6H14O2
Molecular Weight: 118.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 174
Melting point (°C): 43
Flash Point (°C): 77


Other Product Names / Synonyms: 2,3-Dimethyl-2,3-butanediol, Pinacone, Tetramethylethylene Glycol

Formula: C₆H₁₄O₂
Boiling Pt: 174 °C
Melting Pt: 43 °C
Density: 0.967 g/cm³ (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00004462
CAS Number: 76-09-5
EINECS: 200-933-5
Merck Index: 13,07521
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e046a313a51f7a55fcc49d10db413abb3c9d8af8&vid=e046a313a51f7a55fcc49d10db413abb3c9d8af8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR