Propyl-4-hydroxybenzoate ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester

H0219-25G H0219-500G
TCH0219-025GEA 39.83 CAD
TCH0219-025G TCH0219-500G
Propyl-4-hydroxybenzoate ≥99.0% (by titrimetric analysis)
Propyl 4-hydroxybenzoate

CAS Number: 94-13-3
MDL Number: MFCD00002354
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 98
Flash Point (°C): 185


Other Product Names / Synonyms: 4-Hydroxybenzoic Acid Propyl Ester, Propylparaben

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 292
Melting Pt: 98 °C
Density: 1.28 g/cm³ (20 °C)
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00002354
CAS Number: 94-13-3
EINECS: 202-307-7
Merck Index: 13,07958
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ac96bbf572a32a0248564b188b3ebe6aa2bf90f&vid=5ac96bbf572a32a0248564b188b3ebe6aa2bf90f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR