Palmitic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

P0002-500G P0002-25G
TCP0002-500GEA 83.5 CAD
TCP0002-500G TCP0002-025G
Palmitic acid ≥97.0% (by GC, titration analysis)
Palmitic acid

CAS Number: 57-10-3
MDL Number: MFCD00002747
Molecular Formula: C16H32O2
Molecular Weight: 256.43
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 352
Melting point (°C): 64
Flash Point (°C): 171

Formula: C₁₆H₃₂O₂
Boiling Pt: 352 °C
Melting Pt: 64 °C
Density: 0.853 g/cm³ (20 °C)
Flash Pt: 171 °C
Storage Temperature: Ambient
MDL Number: MFCD00002747
CAS Number: 57-10-3
EINECS: 200-312-9
Merck Index: 13,07064
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=783f66713b15eb34c010030c2b868a6d8625908d&vid=79f5297d5cd66c12a5e4c1711456dac9f968ebb1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR