Stretch Nylon Gloves, Protective Industrial Products
Supplier: PIP
98-700 98-701
32932-270CS 3111.07 CAD
32932-270 32932-272
Stretch Nylon Gloves, Protective Industrial Products
Gloves
Lint-free gloves are composed of 100% stretch nylon material that provides limited heat protection.
  • Material is not treated with chemicals of any kind
  • Heavyweight fabric is approximately 120 denier
Launderable, hand-specific gloves are sewn in a full fashion with set-in thumb. Gloves provide dexterity for protecting equipment from human contamination in assembly, examination, and inspection areas.