Dinonyl phthalate (mixture of isomers) ≥95.0% (by GC)
Supplier: TCI America
P0303-500ML
P0303-25ML
TCP0303-500MLEA
671.5
CAD
TCP0303-500ML
TCP0303-025ML
Dinonyl phthalate (mixture of isomers) ≥95.0% (by GC)
Dinonyl phthalate (mixture of isomers)
CAS Number: 84-76-4
MDL Number: MFCD00036237
Molecular Formula: C26H42O4
Molecular Weight: 418.62
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 245
Flash Point (°C): 220
Specific Gravity (20/20): 0.97
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Phthalic Acid Dinonyl Ester
Formula:
C₂₆H₄₂O₄ Boiling Pt: 245 °C Density: 0.97 (20 °C) Flash Pt: 220 °C |
MDL Number:
MFCD00036237 CAS Number: 84-76-4 ADR: 9,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...