Phenol 99.0-100.5% (by anhydrous basis), crystals USP, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = R

MACR0605-04 MACR0605-06
CAMK060504CS 8297.59 CAD
CAMK060504 CAMK060506
Phenol 99.0-100.5% (by anhydrous basis), crystals USP, Macron Fine Chemicals™
Phenol

Crystals.

Formula: C₆H₅OH
Boiling Pt: 181.8 °C (1013 hPa)
Melting Pt: 40.8 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 81 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1,II

Order Now

Specification Test Results

GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C₆H₅OH) (anhydrous basis) 99.0 - 100.5 %
USP - Appearance of Melted Sample Passes Test
USP - Benzene ≤2.0 ppm
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Clarity of Solution and Reaction Passes Test
USP - Congealing Temperature ≥39 °C
USP - Nonvolatile Residue ≤0.05 %
USP - Water (H₂O)(by Karl Fischer titrn) ≤0.5 %
Appearance (colorless to light pink, crystalline mass)
No Class 2 or 3 solvents are expected to be present. The Class 1
solvent Benzene is used as a raw material in a precursor to phenol. The product is tested for any presence of benzene and the result
reported on the Certificate of Analysis.
Product contains about 20 ppm Citric Acid as a preservative.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR